Learn More
E2F4 Monoclonal Antibody (OTI10E7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811690S
Description
E2F4 Monoclonal antibody specifically detects E2F4 in Human samples. It is validated for Western BlotSpecifications
| E2F4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13370 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5140 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI10E7 | |
| Unconjugated | |
| PDE3B | |
| 2010111M04Rik; AI427446; E2F transcription factor 4; E2F transcription factor 4, p107/p130-binding; E2F4; E2F-4; p107/p130-binding protein; transcription factor E2F4 | |
| Human recombinant protein fragment corresponding to amino acids 654-1073 of human PDE3B produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.