missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EAAT2/GLT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59633
This item is not returnable.
View return policy
Description
EAAT2/GLT1 Polyclonal specifically detects EAAT2/GLT1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| EAAT2/GLT1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| GLT1, member 2, solute carrier family 1 (glial high affinity glutamate transporter), member 2, Solute carrier family 1 member 2 | |
| Rabbit | |
| 62 kDa | |
| 100 μL | |
| Hypoxia, Neuroscience, Neurotransmission | |
| 6506 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 2-5 ug/ml | |
| P43004 | |
| SLC1A2 | |
| Synthetic peptides corresponding to SLC1A2(solute carrier family 1 (glial high affinity glutamate transporter), member 2) The peptide sequence was selected from the middle region of SLC1A2. Peptide sequence LVAVDWLLDRMRTSVNVVGDSFGAGIVYHLSKSELDTIDSQHRVHEDIEM The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction