missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ EARS2 293T Cell Transient Overexpression Lysate

Product Code. 18453769 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18453769 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18453769 Supplier Novus Biologicals™ Supplier No. H00124454T01
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Research Category Lipid and Metabolism
Target EARS2
Synonym EC 6.1.1.17, GluRS, Glutamate--tRNA ligase, glutamyl-tRNA synthetase 2, mitochondrial (putative), KIAA1970glutamate tRNA ligase 2, mitochondrial, MSE1, probable glutamyl-tRNA synthetase, mitochondrial
Gene Symbol EARS2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.