Learn More
Abeba™ Easy 37630 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Brand: Abeba™ 37630/38
Additional Details : Weight : 0.97000kg
Description
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Smooth leather, perforated
- White
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3573)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap foldable, adjustable
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Leather | |
Slip-resistant | |
37630 |
White | |
38 | |
Polyurethane |