missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Human Pan-Granulocyte Isolation Kit
Description
- The EasySep™ Human Pan-Granulocyte Enrichment Kit is designed to isolate granulocytes (eosinophils, basophils and neutrophils) from normal peripheral whole blood by negative selection
- Undesired cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-granulocytes and magnetic particles
- The labeled cells are separated using an EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
Specifications
Specifications
| Cell Type | Granulocytes and Subsets |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 Cells |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?