missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Mouse CD4+ T Cell Isolation Kit

Product Code. 15947025
Encompass
Change view
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15947025 1 x 109 cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15947025 Supplier STEMCELL Technologies Supplier No. 19852

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunomagnetic isolation of untouched mouse CD4+ T cells

  • The EasySep™ Mouse CD4+ T Cell Isolation Kit is designed to isolate CD4+ T cells from single-cell suspensions of splenocytes or other tissues by negative selection
  • Unwanted cells are targeted for removal with biotinylated antibodies directed against non-CD4+ T cells and streptavidin-coated magnetic particles (RapidSpheres™ )
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Desired cells are poured off into a new tube
  • This product replaces the EasySep™ Mouse CD4+ T Cell Enrichment Kit (Catalog #19752) for even faster cell isolations

Specifications

Cell Type T Cells, T Cells, CD4+
For Use With (Application) Cell Isolation
Quantity 1 x 109 cells
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.