missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Complete reactor system for organic or aqueous syntheses in the laboratory
- High-quality materials allow operation at temperatures from -50°C to +200°C and up to a vacuum of 3 mbar
Includes
- Ergonomically designed stand system for quick and easy exchange of reactor vessels and adjustment of the working height
- DN 100 (500 mL and 1000 mL) or DN 150 (2000 mL and 5000 mL) vessel clamp for mounting the reactor vessel
- Double-walled reactor vessel made of chemically resistant borosilicate 3.3 glass with bottom discharge valve
- Spring-loaded valve spindle of the drain valve to prevent glass breakage
- Reactor lid with various openings (1 x NS 29/32 center glass joint, 2 x NS 29/32, 1 x NS 14/23) for flexible adaptation to the desired application
- DN 100 or DN 150 clamp ring and FEP-coated O-ring to ensure chemical resistance
- EUROSTAR 60 digital overhead stirrer with 60 Ncm, torque measurement and keyless chuck
- PTFE-coated propeller stirrer for optimum mixing and chemical resistance
- High-quality stirrer guide for vacuum operation up to 3 mbar
- RS 232 and USB interfaces on the stirrer for external control with labworldsoft™ 6 (available as an accessory)
Specifications
Specifications
| Volume (Metric) | 1000 mL |
| Product Type | Lab Reactor |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?