missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EB2 Antibody (KT52), Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP3-23289
This item is not returnable.
View return policy
Description
EB2 Monoclonal antibody specifically detects EB2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| EB2 | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| KT52 | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
| APC-binding protein EB2, EB1, EB2APC-binding protein EB1, End-binding protein 2, microtubule-associated protein, RP/EB family, member 2, RP1microtubule-associated protein RP/EB family member 2, T-cell activation protein, EB1 family | |
| Clone KT52 was generated from a GST fusion protein containing human EB2. This sequence is highly conserved in rat and mouse EB2. [UniProt# Q15555] | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction | |
| 10982 | |
| Store at 4°C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction