missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNB5 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06495PCP
This item is not returnable.
View return policy
Description
GNB5 Polyclonal antibody specifically detects GNB5 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| GNB5 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| FLJ37457, FLJ43714, G protein, beta subunit 5L, G protein, beta-5 subunit, GB5, gbeta5, guanine nucleotide binding protein (G protein), beta 5, guanine nucleotide-binding protein subunit beta-5, guanine nucleotide-binding protein, beta subunit 5L, Transducin beta chain 5 | |
| Produced in rabbits immunized with E. coli-derived Human GNB5 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 10681 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction