missing translation for 'onlineSavingsMsg'
Learn More

Ebrotidine, TRC

Product Code. 30378135
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30378135 500 mg -
30379944 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30378135 Supplier Toronto Research Chemicals Supplier No. TRCE320550500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100981-43-9
Molecular Formula C14H17BrN6O2S3
Molecular Weight (g/mol) 477.42
Synonym [N(E)]-N-[[[2-[[[2-[(Aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]ethyl]amino]methylene]-4-bromobenzenesulfonamide, Ebrodin, FI 3542, Ulsanic
SMILES NC(=N)Nc1nc(CSCCNC=NS(=O)(=O)c2ccc(Br)cc2)cs1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C14H17BrN6O2S3/c15-10-1-3-12(4-2-10)26(22,23)19-9-18-5-6-24-7-11-8-25-14(20-11)21-13(16)17/h1-4,8-9H,5-7H2,(H,18,19)(H4,16,17,20,21)
Formula Weight 475.9758
Chemical Name or Material Ebrotidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.