missing translation for 'onlineSavingsMsg'
Learn More

Ecgonidine Methyl Ester, TRC

Product Code. 30375203
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30375203 1 g -
30369859 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30375203 Supplier Toronto Research Chemicals Supplier No. TRCE3253001G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43021-26-7
Molecular Formula C10 H15 N O2
Molecular Weight (g/mol) 181.23
Synonym Anhydroecgonine Methyl Ester, (1R,5S)-8-Methyl-8-azabicyclo[3.2.1]oct-2-ene-2-carboxylic acid methyl ester, (1R)-8-Methyl-8-azabicyclo[3.2.1]oct-2-ene-2-carboxylic acid methyl ester, (-)-Anhydroecgonine methyl ester, (R)-(-)-Anhydroecgonine methyl ester, EDME, Ecgonidine methyl ester, Methyl ecgonidine
IUPAC Name methyl (1S,5R)-8-methyl-8-azabicyclo[3.2.1]oct-3-ene-4-carboxylate
SMILES COC(=O)C1=CC[C@@H]2CC[C@H]1N2C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H15NO2/c1-11-7-3-5-8(10(12)13-2)9(11)6-4-7/h5,7,9H,3-4,6H2,1-2H3/t7-,9-/m1/s1
Formula Weight 181.1103
Chemical Name or Material Anhydroecgonine methyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.