Learn More
ECHDC3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27098-1-AP-20UL
 View more versions of this product
Product Code. 18044715
  • 145.00€ / 20µL
Description
The ECHDC3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ECHDC3. This antibody recognizes human, canine antigen. The ECHDC3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ECHDC3
150 μg/mL
Rabbit
Primary
Human, Canine
150 kDa
IgG
Western Blot, ELISA
ECHDC3, PP8332
20 μL
79746
Store at -20°C. Stable for one year after shipment.