missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECM-1/Secretory Component P85 Antibody (ECM1/792) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
569.00€
Specifications
| Antigen | ECM-1/Secretory Component P85 |
|---|---|
| Clone | ECM1/792 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Immunofluorescence : 1 - 2 ug/ml, CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF |
Description
ECM-1/Secretory Component P85 Monoclonal specifically detects ECM-1/Secretory Component P85 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence, CyTOF-ready.Specifications
| ECM-1/Secretory Component P85 | |
| 1.0 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF | |
| Unconjugated | |
| Mouse | |
| Extracellular Matrix, Immunology | |
| PBS with No Preservative | |
| 1893 | |
| Recombinant human ECM1 protein | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| ECM1/792 | |
| Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Immunofluorescence : 1 - 2 ug/ml, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| ECM1, extracellular matrix protein 1, Secretory Component Glycoprotein, Secretory component p85 | |
| ECM1 | |
| IgG1 κ | |
| Protein A or G purified | |
| This MAb reacts with a reduction-resistant epitope present in both free and SIgA bound Secretory Component. It does not react with the cell lines lacking secretory component. The antibody is useful for studying the distribution and level of both free and bound secretory component. Secretory component is differentially expressed in epithelium, and the antibody is a popular marker for identifying subpopulations of epithelial cells and epithelial differentiation. The Secretory component antibody is a useful research tool for studying mucosal immunity, inflammation, remodeling, differentiation and tumorigenesis, all processes associated with differential secretory component expression. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title