Learn More
EcoLume LC Fluid, MP Biomedicals
Product Code. 15722252 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 gal.
4 x 1 gal.
Unit Size:
15.14 L
3.79 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15722252 1 gal. 3.79 L
15732252 4 x 1 gal. 15.14 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15722252 Supplier MP Biomedicals Supplier No. 0188247001
  • ONLINE EXCLUSIVE PRICE
    344.00€
    327.60€ / 3.79 L
    Save 16.40€
    4% Off
 Restricted Item
In Stock
Add to basket
Add to basket
Liquid Scintillation Cocktails; EcoLume; MP Biomedical; Biodegradable
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Liquid Scintillation Cocktails
Quantity 1 gal.
compliance-icons
Product Identifier
  • EcoLume Liquid Scintillation Fluid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethoxylated nonylphenol, phenylxylyethane-PXE