Learn More
EcoLume LC Fluid, MP Biomedicals
Product Code. 15732252 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 gal.
4 x 1 gal.
Unit Size:
15.14 L
3.79 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15732252 4 x 1 gal. 15.14 L
15722252 1 gal. 3.79 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15732252 Supplier MP Biomedicals Supplier No. 0188247004
  • ONLINE EXCLUSIVE PRICE
    883.00€
    841.05€ / 15.14 L
    Save 41.95€
    4% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Liquid Scintillation Cocktails; EcoLume; MP Biomedical; Biodegradable
Specifications
Chemical Name or Material Liquid Scintillation Cocktails
Quantity 4 x 1 gal.
compliance-icons compliance-icons
Product Identifier
  • ecolume ES
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Skin sensitiser Category 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: linear alkyl benzene, nonylphenol, ethoxylated, phenylxylyleythane-PXE