Learn More
Econazole Nitrate, TRC
Specifications
Specifications
| Chemical Name or Material | Econazole Nitrate |
| CAS | 24169-02-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18 H15 Cl3 N2 O . H N O3 |
| Quantity | 500 mg |
| Synonym | Econazole Nitrate, 1-[(2RS)-2-[(4-Chlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate, Miconazole Nitrate Imp. B (EP) as Nitrate, (±)-1-[2-[(4-Chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole mononitrate, 1-[2,4-Dichloro-β-[(p-chlorobenzyl)oxy]phenethyl]imidazole mononitrate, (±)-Econazole nitrate, 1-[2-(4-Chlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]imidazole nitrate, Econazole nitrate, Ecostatin, Epi-Pevaryl, Gyno-Pevaril, Gynoryl, Ifenec, Micofugal, Micogin, NSC 243115, Palavale, Pargin, Pevaryl, R 14827, SQ 13050, Spectazole |
| InChI Formula | InChI=1S/C18H15Cl3N2O.HNO3/c19-14-3-1-13(2-4-14)11-24-18(10-23-8-7-22-12-23)16-6-5-15(20)9-17(16)21;2-1(3)4/h1-9,12,18H,10-11H2;(H,2,3,4) |
| SMILES | O[N+](=O)[O-].Clc1ccc(COC(Cn2ccnc2)c3ccc(Cl)cc3Cl)cc1 |
| IUPAC Name | 1-[2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole;nitric acid |
| Molecular Weight (g/mol) | 444.7 |
| Show More |