Description
- Meet ISO Standard 6706.
- Calibrated to contain for measuring final volumes.
- Molded graduations for durability.
- Polypropylene construction provides very good chemical resistance.
Specifications
Specifications
| Capacity (Metric) | 10 mL |
| Material | Polypropylene |
| Certifications/Compliance | ISO 6706 |
| Calibration | To Contain |
| Autoclavable | Autoclavable |
| Shape | Round |
| Base Type | Hexagonal |
| Product Type | Graduated Cylinder |