missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDA/Ectodysplasin Antibody (MM0254-3T2), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12270MFV500
This item is not returnable.
View return policy
Description
EDA/Ectodysplasin Monoclonal antibody specifically detects EDA/Ectodysplasin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| EDA/Ectodysplasin | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 1896 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0254-3T2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ectodysplasin A, ED1 | |
| Human recombinant EDA/Ectodysplasin (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction