missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDA Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25892-1-AP-150UL
This item is not returnable.
View return policy
Description
EDA is a type II membrane protein that can be cleaved by furin to produce a secreted form. This protein, which belongs to the tumor necrosis factor family, acts as a homotrimer and may be involved in cell-cell signaling during the development of ectodermal organs. Defects in the gene for EDA are a cause of ectodermal dysplasia, anhidrotic, which is also known as X-linked hypohidrotic ectodermal dysplasia.Specifications
| EDA | |
| Polyclonal | |
| Unconjugated | |
| EDA | |
| Ectodermal dysplasia protein, ectodysplasin A, ED1, ED1 A1, ED1 A2, EDA, EDA protein, EDA1, EDA2, HED, XHED, XLHED | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13607, 1896 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O54693, Q92838 | |
| EDA | |
| EDA Fusion Protein Ag23154 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction