missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16486-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that is important in mRNA degradation. The encoded protein is a component of a decapping complex that promotes efficient removal of the monomethylguanosine (m7G) cap from mRNAs, as part of the 5′ to 3′ mRNA decay pathway. Mutations in this gene have been identified in human patients with an autosomal recessive form of intellectual disability.Specifications
| EDC3 | |
| Polyclonal | |
| Unconjugated | |
| EDC3 | |
| EDC3, hYjeF_N2, LSM16, LSM16 homolog, YJDC, YjeF_N2, YJEFN2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 315708, 353190, 80153 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8K2D3, Q96F86 | |
| Edc3 | |
| EDC3 Fusion Protein Ag9608 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction