missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEA1 Rabbit anti-Human, Mouse, Rat, Clone: 2C11, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 81245-1-RR-100UL
This item is not returnable.
View return policy
Description
EEA1 Recombinant Monoclonal antibody specifically detects EEA1 in Human, Mouse, Rat samples. It is validated for WB, ELISASpecifications
| EEA1 | |
| Recombinant Monoclonal | |
| 1000 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| Rabbit | |
| 100 μL | |
| 8411 | |
| Store at -20°C | |
| Liquid |
| Western Blot, ELISA | |
| 2C11 | |
| Unconjugated | |
| BC002409 | |
| Protein A purification | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction