missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF1A1 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
129.00€ - 399.00€
Specifications
| Antigen | EEF1A1 |
|---|---|
| Clone | 2B16 |
| Concentration | 250 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235812
|
Proteintech
81377-1-RR-100UL |
100 μL |
439.00€ 399.00€ / 100µL Save 40.00€ 9% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235135
|
Proteintech
81377-1-RR-20UL |
20 μL |
149.00€ 129.00€ / 20µL Save 20.00€ 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The EEF1A1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human EEF1A1. This antibody recognizes human, mouse, rat antigen. The EEF1A1 antibody has been validated for the following applications: WB, IHC, IF/ICC, IP, ELISA analysis.Specifications
| EEF1A1 | |
| 250 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| 1915 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 2B16 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| CCS-3; CCS3; EEF-1; EEF1A; EF-Tu; EF1A; FLJ25721; GRAF-1EF; HNGC:16303; LENG7; MGC102687; MGC131894; MGC16224; PTI1; eEF1A-1 | |
| IgG | |
| Protein A purification | |
| Recombinant Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title