Learn More
EEF1A2 Mouse anti-Human, Unconjugated, Clone: 4F3F4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66806-1-IG-150UL
Description
The EEF1A2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EEF1A2. This antibody recognizes human, mouse, rat, pig antigen. The EEF1A2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| EEF1A2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot | |
| 4F3F4 | |
| Unconjugated | |
| 150 μL | |
| 1917 | |
| IgG1 |