Learn More
EEF1A2 Mouse anti-Human, Unconjugated, Clone: 4F3F4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66806-1-IG-150UL
 View more versions of this product
Product Code. 16899015
  • 410.00€ / 150µL
Description
The EEF1A2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EEF1A2. This antibody recognizes human, mouse, rat, pig antigen. The EEF1A2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
EEF1A2
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot
4F3F4
Unconjugated
150 μL
1917