Learn More
EFHC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11471-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes an EF-hand-containing calcium binding protein. The encoded protein likely plays a role in calcium homeostasis. Mutations in this gene have been associated with susceptibility to juvenile myoclonic epilepsy and juvenile absence epilepsy. Alternatively spliced transcript variants have been described.Specifications
| EFHC1 | |
| Polyclonal | |
| Unconjugated | |
| EFHC1 | |
| EFHC1, EJM, EJM1, JAE, Myoclonin 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114327, 301295, 71877 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5JVL4, Q9D9T8 | |
| Efhc1 | |
| EFHC1 Fusion Protein Ag2048 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |