Learn More
EGFR Antibody (C225 (Cetuximab)) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-52671-0.025MG
This item is not returnable.
View return policy
Description
EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, CyTOF-ready, Block/Neutralize.
Specifications
| EGFR | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
| EGFR | |
| The epidermal growth factor receptor is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands. | |
| 0.025 mg | |
| Cancer, Cell Biology, Cell Cycle and Replication, Growth and Development, Hypoxia, Signal Transduction, Tumor Suppressors, Tyrosine Kinases | |
| 1956 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Blocking Assay, CyTOF | |
| C225 (Cetuximab) | |
| Flow Cytometry 1:10 - 1:1000, ELISA 1:100-1:2000, Blocking/Neutralizing 50 - 200 molar excess, CyTOF-ready | |
| avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognises human EGFR. | |
| Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. | |
| IgG |