missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGFR Antibody (GFR/1708), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54330AF647
This item is not returnable.
View return policy
Description
EGFR Monoclonal antibody specifically detects EGFR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| EGFR | |
| Monoclonal | |
| Alexa Fluor 647 | |
| avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
| Recombinant full-length human EGFR protein (Uniprot: P00533) | |
| 100 μg | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Cellular Signaling, Growth and Development, Hypoxia, Signal Transduction, Tumor Suppressors, Tyrosine Kinases | |
| 1956 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| GFR/1708 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction