missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGLN1/PHD2 Antibody (366G/76/3), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08507AF700
This item is not returnable.
View return policy
Description
EGLN1/PHD2 Monoclonal antibody specifically detects EGLN1/PHD2 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| EGLN1/PHD2 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| C1orf12, EC 1.14.11.29, EC:1.14.11.291, ECYT3, egl nine homolog 1, egl nine-like protein 1, EGLN1, HALAH, HIF prolyl hydroxylase 2, HIFPH2, HIF-PH2, HIF-prolyl hydroxylase 2, HPH2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2, PNAS-118, PNAS-137, Prolyl hydroxylase domain-containing protein 2, SM20, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6 | |
| Residues 1-24 of EGLN1/PHD2 (Uniprot: Q9GZT9) | |
| 100 μg | |
| Angiogenesis, Cancer, Chromatin Research, Hypoxia, Immunology, Lipid and Metabolism | |
| 54583 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 366G/76/3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction