missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGLN3/PHD3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-303SS
This item is not returnable.
View return policy
Description
EGLN3/PHD3 Polyclonal specifically detects EGLN3/PHD3 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting.
Specifications
| EGLN3/PHD3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| EC 1.14.11, EC 1.14.11.-, EGL nine (C.elegans) homolog 3, egl nine homolog 3, egl nine homolog 3 (C. elegans), egl nine-like protein 3 isoform, FLJ21620, HIF prolyl hydroxylase 3, HIF-PH3, HIFPH3 MGC125998, HIF-prolyl hydroxylase 3, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 3, MGC125999, pdh3, PHD3, Prolyl hydroxylase domain-containing protein 3 | |
| Rabbit | |
| 27 kDa | |
| 0.025 mL | |
| Apoptosis, Cancer, Hypoxia | |
| 112399 | |
| Human, Mouse, Primate | |
| IgG |
| Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Simple Western 1:1000, Chromatin Immunoprecipitation reported in scientific literature (PMID 21620138), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 20801873), Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 15721254), Immunohistochemistry-Paraffin reported in scientific literature (PMID 25161887), Immunoblotting, Chromatin Immunoprecipitation (ChIP) | |
| Q9H6Z9 | |
| EGLN3 | |
| A synthetic peptide made to the C-terminus of humanPHD3/HIF Prolyl Hydroxylase 3. [LocusLink ID 112399] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and mouse. Primate reactivity reported in scientific literature (PMID: 23732909). mmunogen sequence has 92% homology to rat. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction