Learn More
Lonza™ EGM™ Endothelial Cell Growth Medium SingleQuots™ Kit
Product Code. 11685490
Change view
Click to view available options
hEGF: 0.5 mL, Insulin: 0.5 mL, Hydrocortisone: 0.5 mL, GA-1000: 0.5 mL, BBE: 2.0 mL, Ascorbic Acid: 0.5 mL, FBS: 10.0 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11685490 hEGF: 0.5 mL, Insulin: 0.5 mL, Hydrocortisone: 0.5 mL, GA-1000: 0.5 mL, BBE: 2.0 mL, Ascorbic Acid: 0.5 mL, FBS: 10.0 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11685490 Supplier Lonza™ Supplier No. CC4133
Supplements and growth factors kit for use with endothelial cell basal medium.
  • Separately sold SingleQuot™ kit for supplementing basal medium.
  • Part of the full EGM™ Endothelial Cell Growth Medium BulletKit™ (catalog number CC-3124), which consists of basal medium and supplements packaged together.
  • Includes the following:
    • 1 x Green Cap Vial with hEGF 0.50 mL
    • 1 x Natural Cap Vial with Hydrocortisone 0.50 mL
    • 1 x Red Cap Vial with GA-1000 0.50 mL
    • 1 Bottle BBE 2.00 mL
    • 1 x Blue Cap Vial with Ascorbic Acid 0.50 mL
    • 1 Bottle FBS 10.00 mL
TRUSTED_SUSTAINABILITY
Specifications
Cell Type Endothelial Cells
Product Type Supplements and growth factors
Quantity hEGF: 0.5 mL, Insulin: 0.5 mL, Hydrocortisone: 0.5 mL, GA-1000: 0.5 mL, BBE: 2.0 mL, Ascorbic Acid: 0.5 mL, FBS: 10.0 mL
Content And Storage hEGF, Hydrocortisone, GA-1000, BBE, Ascorbic Acid, FBS