missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine IL-12/IL-23 p40 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1969-SP
This item is not returnable.
View return policy
Description
IL-12/IL-23 p40 Polyclonal specifically detects IL-12/IL-23 p40 in Canine samples. It is validated for Western Blot, Neutralization.
Specifications
| IL-12/IL-23 p40 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL12 p40, IL-12 subunit p40, IL12B, IL-12B, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2, NKSF2IL12, subunit p40 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 403976 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.1-0.4 ug/mL | |
| Q28268 | |
| IL12B | |
| Mouse myeloma cell line NS0-derived recombinant canine IL-12/IL-23 p40 Ile23-Ser329 Accession # Q28268 | |
| 25 μg | |
| Primary | |
| Detects canine IL-12/IL-23 p40 in direct ELISAs and Western blots. | |
| Canine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction