missing translation for 'onlineSavingsMsg'
Learn More

Eicosapentaenoic Acid, TRC

Product Code. 30388056
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388056 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388056 Supplier Toronto Research Chemicals Supplier No. TRCE477800250MG
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material cis Eicosapentaenoic acid (EPA)
CAS 10417-94-4
Molecular Formula C20 H30 O2
Quantity 250 mg
Synonym 5,8,11,14,17-Eicosapentaenoic acid, (5Z,8Z,11Z,14Z,17Z)-, 5,8,11,14,17-Eicosapentaenoic acid (6CI), 5,8,11,14,17-Eicosapentaenoic acid, (all-Z)- (8CI), (5Z,8Z,11Z,14Z,17Z)-5,8,11,14,17-Eicosapentaenoic acid, (5Z,8Z,11Z,14Z,17Z)-Eicosa-5,8,11,14,17-pentaenoic acid, (5Z,8Z,11Z,14Z,17Z)-Eicosapentaenoic acid, (all-Z)-5,8,11,14,17-Eicosapentaenoic acid, (all-Z)-δ5,8,11,14,17-Eicosapentaenoic acid, (all-cis)-5,8,11,14,17-Eicosapentaenoic acid, EPA, EPA 45G, Eicosapentaenoic acid, Eye-Q, Icosapent, Incromega E 7010SR, PlusEPA, Ropufa 70, Timnodonic acid, cis-5,8,11,14,17-Eicosapentaenoic acid, o 3Mega+ Joy
InChI Formula InChI=1S/C20H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h3-4,6-7,9-10,12-13,15-16H,2,5,8,11,14,17-19H2,1H3,(H,21,22)/b4-3-,7-6-,10-9-,13-12-,16-15-
SMILES CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)O
IUPAC Name (5Z,8Z,11Z,14Z,17Z)-icosa-5,8,11,14,17-pentaenoic acid
Molecular Weight (g/mol) 302.45
Formula Weight 302.2246
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.