missing translation for 'onlineSavingsMsg'
Learn More

eIF2 alpha/EIF2S1, p Ser51 Antibody (SR1604), Novus Biologicals™

Rabbit Monoclonal Antibody

349.00€ - 472.00€

Specifications

Antigen eIF2 alpha/EIF2S1
Clone SR1604
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:40, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18663474
View Documents
Novus Biologicals
NBP3-21506-100ul
100 μg
472.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18601024
View Documents
Novus Biologicals
NBP3-21506-50ul
50 μg
349.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

eIF2 alpha/EIF2S1 Monoclonal antibody specifically detects eIF2 alpha/EIF2S1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Specifications

Specifications

eIF2 alpha/EIF2S1
Western Blot 1:500-1:2000, Flow Cytometry 1:40, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Monoclonal
Purified
RUO
Human, Mouse, Rat
EIF2, EIF-2, EIF2A, EIF-2A, EIF-2alpha, EIF2S1 eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa
A synthesized peptide derived from human phospho-eIF2 alpha/EIF2S1 [Ser51] (Uniprot #: P05198)
Primary
Store at -20 degrees C. Avoid freeze/thaw cycles.
SR1604
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Unconjugated
Rabbit
Chromatin Research, DNA Repair, Lipid and Metabolism, Phospho Specific, Translation Control, Unfolded Protein Response
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
1965
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.