missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 405 Anti-Mouse CD11a (M17/4), Proteintech
Rat Monoclonal Antibody
Brand: Proteintech CL405-65177-100UG
This item is not returnable.
View return policy
Description
The CD11a antibody from Proteintech is a rat monoclonal antibody generated with c57bl/6 mouse splenic secondary cytotoxic t cells of mouse CD11a. This antibody recognizes mouse antigen. The CD11a antibody has been validated for the following applications: FC analysis. This antibody is coralite™ plus 405.Specifications
| CD11a | |
| Monoclonal | |
| 0.5 mg/mL | |
| Rat | |
| Purified by protein-A affinity chromatography | |
| RUO | |
| 16408 | |
| Store at 2-8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry | |
| M17/4 | |
| CoraLite Plus 405 | |
| C57BL/6 mouse splenic secondary cytotoxic T cells | |
| 100 μg | |
| Primary | |
| Mouse | |
| Flow Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction