Learn More
EIF3E Mouse anti-Human, Unconjugated, Clone: 1C6C8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67095-1-IG-20UL
Description
The EIF3E antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EIF3E. This antibody recognizes human, mouse, rat antigen. The EIF3E antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
| EIF3E | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunoprecipitation, ELISA | |
| 1C6C8 | |
| Unconjugated | |
| 20 μL | |
| 3646 | |
| IgG1 |