missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4B, p Ser406 Antibody (SR2202), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21605-50ul
This item is not returnable.
View return policy
Description
eIF4B Monoclonal antibody specifically detects eIF4B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| eIF4B | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR2202 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| EIF-4B, eukaryotic translation initiation factor 4B, PRO1843 | |
| A synthesized peptide derived from human phospho-eIF4B [Ser406] (Uniprot #: P23588) | |
| 50 μg | |
| Core ESC Like Genes, mTOR Pathway, Phospho Specific, Stem Cell Markers | |
| 1975 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction