Learn More
EIF4G1 Mouse anti-Human, Unconjugated, Clone: 2B10G8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67199-1-IG-150UL
Description
The EIF4G1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EIF4G1. This antibody recognizes human, mouse, rat antigen. The EIF4G1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| EIF4G1 | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunofluorescence, ELISA | |
| 2B10G8 | |
| Unconjugated | |
| 150 μL | |
| 1981 | |
| IgG1 |