Learn More
Invitrogen™ EK-Away™ Resin
Product Code. 10403982
Change view
Click to view available options
Quantity:
7.5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10403982 7.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10403982 Supplier Invitrogen™ Supplier No. R18001
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes
EK-Away is provided as a 50% ethanol slurry; 10X binding and 10X stripping buffers are included; 7.5mL resin will remove 250 units of enterokinase; 30mL will remove 1000 units of EKMax.
Specifically designed for removal of EKMaxor other enterokinase enzymes following cleavage of proteins containing the enterokinase cleavage site
  • The resin is conjugated with soybean trypsin inhibitor, which has a high affinity and binding capacity for enterokinase
  • The enzyme catalytic site binds this agarose-based resin for simple batch or column-bound removal of EKMaxor other enterokinase preparations
GST-Tagged Protein Purification, His-Tagged Protein Purification, Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage EK-Away™ is provided as a 50% ethanol slurry and should be stored at +4°C. 10X binding and 10X stripping buffers are included. 7.5 ml resin will remove 250 units of enterokinase; 30 ml will remove 1000 units of EKMax™. All reagents are guaranteed stable for 6 months when stored properly.
Form Liquid Suspension
Product Line EK-Away
Quantity 7.5 mL
Type Resin
Stationary Phase Enterokinase
Column Type Affinity