missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PL BioScience ELAREM™ Perform-FD PLUS, Fibrinogen-depleted Human Platelet Lysate, No Heparin Added or Required
missing translation for 'orderingAttributeHoverText'
Quantity:
50 mL
100 mL
500 mL
missing translation for 'unitSize'
100mL
500mL
50mL
Beschreibung
- Clot-free and Fibrinogen-depleted HPL: ELAREM™ Perform-FD PLUS is a fibrinogen-depleted, ready-to-use Human Platelet Lysate that is widely applicable.
- No Heparin Added or Required: Due to the unique fibrinogen-depletion technique, ELAREM™ Perform-FD PLUS is clot-free and heparin-free. The product neither requires the addition of anticoagulants (e.g. heparin) in cell culture nor during manufacturing.
- High Performance Lab Process: ELAREM™ Perform-FD PLUS efficiently and safely supports in vitro expansion of various primary cells and cell lines.
- EU Origin for Long Traceability: ELAREM™ Perform-FD PLUS is manufactured from platelet units obtained from healthy blood donors at licensed blood centers in the EU. ELAREM™ Perform-FD PLUS is derived from EU origin platelet units which ensures long raw material traceability periods.
- Intended Use: ELAREM™ Perform-FD PLUS is for in vitro and manufacturing use only. The product is not intended for direct therapeutic use. For safety precautions, please see appropriate Safety Data Sheet (SDS).
Spezifikation
Spezifikation
| Color | Yellow |
| Culture Type | Adherent Cell Culture, Suspension Cell Culture |
| Form | Liquid |
| Product Type | Cell Culture Supplement |
| Sterility | Sterile |
| Sterilization Method | Sterile-filtered |
| Formulation | Liquid, Human Platelet Lysate, Fibrinogen-depleted |
| Purity or Quality Grade | Research Grade |
| Quantity | 100 mL |
| Shipping Condition | Dry Ice |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur