missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elastase Antibody (CELA3B/1811), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54489B
This item is not returnable.
View return policy
Description
Elastase Monoclonal antibody specifically detects Elastase in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF
Specifications
| Elastase | |
| Monoclonal | |
| Biotin | |
| CBPP, cholesterol-binding pancreatic protease, chymotrypsin-like elastase family member 3B, chymotrypsin-like elastase family, member 3B, E1, EC 3.4.21, EC 3.4.21.70, EL-1, ELA3B, elastase 1, elastase 3B, pancreatic, Elastase IIIB, elastase-3B, fecal elastase 1, pancreatic elastase 1, pancreatic endopeptidase E, Protease E, proteinase E | |
| Recombinant human Elastase protein fragment (around aa 82-238) (exact sequence is proprietary) (Uniprot: P08861) | |
| 100 μg | |
| Cancer, Neuroscience | |
| 23436 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| CELA3B/1811 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction