missing translation for 'onlineSavingsMsg'
Learn More

Electrode Regenerative Solution, Chem-Lab

Product Code. 11957755
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11957755 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11957755 Supplier Chem-Lab Supplier No. CL0218370100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Restore an electrode that is starting to perform poorly. Electrode Regenerative Solution, Chem-Lab is an ammonium hydrogen difluoride solution that can help to regenerate the pH-sensitive glass membrane of an electrode.

Over time, electrodes may start displaying excessive response time, even when they have been carefully handled and properly stored. In these cases, it may be possible to reactivate the electrode by immersing it in an ammonium bifluoride regeneration solution. The solution works by etching away a very thin layer of the glass membrane, exposing a fresh surface area.

Specifications

CAS 1341-49-7
Boiling Point 100°C
Color Colorless
Density 1.00g/mL
For Use With (Equipment) Glass Electrodes
Physical Form Liquid
Quantity 100 mL
Molecular Formula F2H5N
Odor Odorless
MDL Number MFCD00012018
Packaging Bottle
Solubility Information Fully miscible
InChI Key KVBCYCWRDBDGBG-UHFFFAOYSA-N
SMILES N.F.F
IUPAC Name amine dihydrofluoride
Molecular Weight (g/mol) 57.04
Formula Weight 57.04g/mol
Chemical Name or Material Ammonium hydrogen difouride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Regeneration solution for electrodes
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.