missing translation for 'onlineSavingsMsg'
Learn More

Ebro Electronic™ TPX 330 Probe

Product Code. 10717093
Change view
Click to view available options
Temperature Range (Metric):
-100°C to 500°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) unitSize
10717093 -100°C to 500°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10717093 Supplier Ebro Electronic™ Supplier No. 13415439
Add to basket
Add to basket
This item is not returnable. View return policy

Probe for TFX 430/GFX 460

PT 100, 4 conductors class A, Lemo size 1
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012; EN 13485
Diameter (Metric) Probe 8 mm
Disposable Single-use
For Use With (Application) Applications: Laboratory, temperature monitoring, measure air, fluids, ductile substances, aggressive media and surfaces, Process monitoring
Height (Metric) 54 mm
Immersion Type Total or Partial
Length (Metric) 355 mm
Material Stainless Steel, Plastic
Model TFX 430 & TPX 330
Probe Type PT 100 class protected
Resolution 0.01°C from −10.00°C to 199.99°C, 0.1°C remaining range
Weight (Metric) 0.4 kg
Product Type Probe
Temperature Range (Metric) -100°C to 500°C
For Use With (Equipment) AG 130, AG 140, AG 150, AX 110, TPX 130, TPX 230, TPX 330, TFX 430
Width (Metric) 162 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.