Learn More
Fisherbrand™ PTFE-Coated Rare Earth Core Stir Bars, Elliptical
Product Code. 16235801 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Length (Metric):
10 mm
15 mm
25 mm
70 mm
Diameter (Metric):
6 mm
10 mm
14 mm
28 mm
Unit Size:
Each
Pack of 10
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Unit Size
16235801 10 mm 6 mm Pack of 10
16245801 15 mm 10 mm Pack of 10
11848922 25 mm 14 mm Pack of 5
10650693 70 mm 28 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16235801
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Use for significantly increased magnetic strength and resistance to demagnetization. Rare earth stir bars are easily identified by a chemically inert carbon black spot.
  • Use rare earth magnets for significantly increased magnetic strength compared to those using AlNiCo magnets
  • Samarium-cobalt core is resistant to demagnetization
  • Identified by a chemically inert, carbon black spot
The high magnetic coupling power of rare earth stir bars may cause abrasion of the PTFE coating against the base of the container. When stirring arrays are used, as in multi-well plates, lateral interaction between stir bars may be observed.
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Samarium-Cobalt magnet core
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Diameter (Metric) 6 mm
Length (Metric) 10 mm
Quantity 2/Pk.
Shape Elliptical