Learn More
Tuttnauer ELV-D Vertical Autoclave
Product Code. 17878988 Shop All Tuttnauer Products
Change view
Click to view available options
Capacity (Metric):
31 L
65 L
85 L
160 L
Features:
Single Phase
Single Phase, Water Reservoir
Three Phase
Three Phase, BioHazard Filter
Three Phase, Water Reservoir
Water Reservoir
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Features Unit Size
17878988 160 L Single Phase Each
17801077 31 L Water Reservoir Each
17871087 65 L Single Phase, Water Reservoir Each
17811077 65 L Three Phase Each
17801097 85 L Water Reservoir Each
17891087 85 L Single Phase, Water Reservoir Each
17818998 160 L Single Phase, Water Reservoir Each
17868988 160 L Three Phase, Water Reservoir Each
17808998 160 L Three Phase Each
17828998 160 L Three Phase, BioHazard Filter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17878988 Supplier Tuttnauer Supplier No. 5075ELVCPVGP230D
  • ONLINE EXCLUSIVE PRICE
    37915.50€
    33097.75€ / Pack of 1
    Save 4817.75€
    12% Off
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
The ELV-D Vertical Autoclave has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, biohazard and waste sterilization, Fo control, and more.
Designed for your laboratory applications. The 5075ELV-D has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, Fo control, and more.
  • Advanced Control System - Take Advantage of Tuttnauer's state-of-the-art control system with multi-color display
  • Safe - Automatic pressure locking system. Door will not open when the chamber is overpressured
  • Optional Internal Printing - Documents date, time, temperature and pressure
  • Intuitive Multi-Color Display - Multi-color display for easier reading gives quick access to important information. Color is used to indicate the stage of the cycle
  • High Quality Stainless Steel - 316L grade stainless steel chamber and door with electro-polish finish
  • Diverse Load Types - The 5075ELV-D autoclave comes with 8 factory set cycle programs that cover the processing needs of most laboratories and research centers
Specifications
Capacity (English) 35.2 gal.
Capacity (Metric) 160 L
Depth (Metric) Chamber 75 cm
Diameter (English) Chamber 20 in.
Depth (English) Chamber 30 in.
Diameter (Metric) Chamber 50 cm
Length (English) 30.8 in.
Length (Metric) 77 cm
Model 5075ELV-D
Product Type Autoclave, Top-loading
Temperature (English) 221°F to 280°F
Temperature (Metric) 105°C to 138°C
Features Single Phase
Includes Cooling, Vacuum, Steam Generator, Printer