missing translation for 'onlineSavingsMsg'
Learn More

Merck Accessories, and Replacement Parts for In-Line Filter Holder

Accessory and replacements for 47mm In-Line Filter Holder

Brand:  Merck XX4304704

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10089970

  • 59.15€ / Pack of 2

Estimated Shipment: 23-05-2023
to see stock.
Add to basket

Description

Description

Accessory and replacements for 47mm In-Line Filter Holder

Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.Tri-Clamp (TC) fittings are referred to by convention as 1 1/2 in., 2 in., etc. 1 1/2 in. TC fittings connect to 1 in. or 1 1/2 in. O.D. tube sizes. 2 in. TC fittings connect to 2 in. O.D. tube size only.
Specifications

Specifications

Not Autoclavable
47 mm In-line filter holder
Non-sterile
1/4 in.
6.3 mm
Hose Adapter
Polypropylene
Hose adapter, 1/4 in. NPTF to 5/16 in. ID, polypropylene
6 mm
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions