missing translation for 'onlineSavingsMsg'
Learn More

Merck pET-41b(+) DNA Vector

Product Code. 17747668
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17747668 10 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17747668 Supplier Merck Supplier No. 705573
  • ONLINE EXCLUSIVE PRICE
    507.00€
    481.43€ / Pack of 1
    Save 25.57€
    5% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for cloning and high-level expression of peptide sequences fused with the 220 aa GST·Tag™ protein.

The sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map. The f1 origin is oriented so that infection with helper phage will produce virions containing single stranded DNA that corresponds to the coding strand. Vector encoded sequence can be completely removed when cloning into the PshAI site (as shown below) and then cleaving the GST fusion protein with Enterokinase.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -70°C. Avoid freeze/thaw cycles
Quantity 10 μg

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.