Learn More
Merck Ultrafree™-DA Centrifugal Filter
Description
Includes
50 gel nebulizers, 50 Ultrafree-MC 0.45μm separators, 50 microcentrifuge vials
Recover 100 to 10,000 bp DNA from agarose gel slices in a single 10-minute spin. The kit consists of a pre-assembled filter device with an agarose gel nebulizer, a microcentrifuge vial, and modified TAE gel extraction buffer (dry).The device utilizes gel compression to extract DNA from the agarose. Centrifugal force collapses the gel structure, drives the agarose through a small orifice in the gel nebulizer and the resultant gel slurry is sprayed into the sample filter cup.Prepared DNA requires no further purification for most applications, including cloning and radioisotopic or fluorescent DNA sequencing. Since agarose gel electrophoresis has high resolving power, the small and large non-specific amplification products that frequently interfere with cloning and sequencing after PCR are completely removed from the product.
Typical DNA Recoveries from Agarose Gels
Protocol
1. Excise the band of interest after routine electrophoretic analysis of PCR products or other DNA through an agarose gel.
2. Place the gel slice into the pre-assembled device. Note: Protocol is not compatible with low melting point agarose.
3. Spin at 5,000 x g for 10 minutes to extract gel slurry into filter cup.
4. DNA can be stored in the capped filtrate vial after discarding the sample filter cup and gel nebulizer.
- Minimal hands-on time
- 10-minute spin
- Fully-functional DNA
Typical DNA Recoveries from Agarose Gels
| DNA Size (bp) | Percent DNA Recoveries |
| 100 | 78 |
| 700 | 71 |
| 1000 | 77 |
| 2027 | 47 |
| 4361 | 35 |
| 9416 | 32 |
| 23130 | 29 |
Protocol
1. Excise the band of interest after routine electrophoretic analysis of PCR products or other DNA through an agarose gel.
2. Place the gel slice into the pre-assembled device. Note: Protocol is not compatible with low melting point agarose.
3. Spin at 5,000 x g for 10 minutes to extract gel slurry into filter cup.
4. DNA can be stored in the capped filtrate vial after discarding the sample filter cup and gel nebulizer.
An Amicon™ Bioseparations product.
Specifications
Specifications
| For Use With (Application) | DNA Gel Extraction |
| Includes | 50 Gel nebulizers, 50 Ultrafree™-MC 0.45 μm separators, 50 microcentrifuge vials |
| Product Type | Accessory |
| Quantity | 50 Pack |