missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EMILIN2 Monoclonal Antibody (1031401)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA560897
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
EMILIN2 is a protein coding gene. May be responsible for anchoring smooth muscle cells to elastic fibers, and may be involved not only in the formation of the elastic fiber, but also in the processes that regulate vessel assembly. Has cell adhesive capacity.
Specifications
| EMILIN2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9BXX0 | |
| EMILIN2 | |
| Human EMILIN2 synthetic peptide. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunocytochemistry | |
| 1031401 | |
| Unconjugated | |
| EMILIN2 | |
| elastin microfibril interface-located protein 2; Elastin microfibril interfacer 2; EMILIN2; EMILIN-2; extracellular glycoprotein EMILIN-2; FLJ33200; FOAP-10; Protein FOAP-10 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 84034 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction