Learn More
Syngene™ Emission Filter
Syngene™ Emission Filter is for use in Syngene Gbox and Gbox Mini Systems 7 position filter wheel
Brand: Syngene™ FILT585-GX
Additional Details : Weight : 0.50000kg
Description
- For use with Gbox and Gbox Mini Systems 7 position filter wheel
Specifications
1 | |
585 nm | |
Filter |
Gbox and Gbox Mini | |
7 Position Filter Wheel, Multiplex Suitable |