missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EMP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567570
This item is not returnable.
View return policy
Description
EMP1 Polyclonal Antibody for IHC (P)
EMP1 is a protein coding gene.
Specifications
| EMP1 | |
| Polyclonal | |
| Unconjugated | |
| EMP1 | |
| B4B; CL-20; Emp1; EMP-1; ENP1MR; Epithelial membrane protein 1; I-8-09; Protein B4B; TMP; tumor-associated membrane protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2012 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P54849 | |
| EMP1 | |
| Synthesized peptide derived from human EMP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction