Learn More
Buerkle™ Emptying Device for Zone Sampler
Buerkle™ Emptying Device for Zone Sampler allows samples to be emptied directly into the sample containers.
Brand: Buerkle™ 5317-0030
497.91 EUR valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Additional Details : Weight : 3.03550kg
Description
- Saves valuable time
- Reduces contamination
- Individual funnels can be set to the different axis distances of the collection chambers
Specifications
Emptying Device | |
3 Funnels | |
100 cm |
Stainless Steel (V2A) | |
Zone Samplers |